siRNA Details
| Gene Symbol: Zfp960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC018101.1 | - | 2664 | 3494875 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp960| Gene Symbol: Zfp960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC018101.1 | - | 2664 | 3494875 | ||