siRNA Details
| Gene Symbol: Gm13084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005371.3 | NP_001005371.2 | 2 | 136 | ||
| GenBank mRNA | AK145369.1 | - | 2 | 133 | ||
| BC080785.1 | - | 2 | 134 | 30481291 | ||
| BY029847.1 | - | 2 | 135 | |||
| CJ082474.1 | - | 2 | 137 | |||

Gm13084
Genomic Map