siRNA Details
| Gene Symbol: Gm13084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005371.3 | NP_001005371.2 | 3, 4 | 947 | ||
| GenBank mRNA | AK145369.1 | - | 3, 4 | 944 | ||
| BC080785.1 | - | 3, 4 | 945 | 30481291 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13084| Gene Symbol: Gm13084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005371.3 | NP_001005371.2 | 3, 4 | 947 | ||
| GenBank mRNA | AK145369.1 | - | 3, 4 | 944 | ||
| BC080785.1 | - | 3, 4 | 945 | 30481291 | ||