siRNA Details
| Gene Symbol: Ddx19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005381.2 | NP_001005381.2 | 6, 7 | 553 | ||
| XM_008772490.1 | XP_008770712.1 | 339 | ||||
| GenBank mRNA | AY303540.1 | - | 571 | |||
| BC079094.1 | - | 6, 7 | 524 | 7123989 | ||
| CB582878.1 | - | 6, 7 | 553 | |||

Ddx19a
Genomic Map