siRNA Details
| Gene Symbol: KRTAP5-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005405.2 | NP_001005405.1 | 1 | 722 | ||
| GenBank mRNA | AB126080.1 | - | 1 | 722 | ||
| AJ628244.1 | - | 1 | 144 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP5-11| Gene Symbol: KRTAP5-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005405.2 | NP_001005405.1 | 1 | 722 | ||
| GenBank mRNA | AB126080.1 | - | 1 | 722 | ||
| AJ628244.1 | - | 1 | 144 | |||