siRNA Details
| Gene Symbol: KRBOX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129898.1 | NP_001123370.1 | 4 | 408 | ||
| NM_001129899.1 | NP_001123371.1 | 4 | 408 | |||
| NM_001129900.1 | NP_001123372.1 | 4 | 408 | |||
| NM_017776.2 | NP_060246.2 | 4 | 408 | |||
| GenBank mRNA | AK000351.1 | - | 4 | 273 | ||
| AK096413.1 | - | 4 | 300 | |||
| AK289703.1 | - | 4 | 305 | |||
| BC012569.1 | - | 3 | 219 | 4283420 | ||
| DA152034.1 | - | 4 | 339 | |||
| DA631861.1 | - | 4 | 325 | |||
| DA877282.1 | - | 4 | 409 | |||
| Gene Symbol: ZNF674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039891.2 | NP_001034980.1 | 4 | 336 | ||
| NM_001146291.1 | NP_001139763.1 | 4 | 336 | |||
| NM_001190417.1 | NP_001177346.1 | 4 | 336 | |||
| XM_011543941.2 | XP_011542243.1 | 4 | 333 | |||
| XM_011543943.2 | XP_011542245.1 | 4 | 338 | |||
| XM_011543944.2 | XP_011542246.1 | 2 | 143 | |||
| XM_017029728.1 | XP_016885217.1 | 4 | 299 | |||
| XM_017029729.1 | XP_016885218.1 | 4 | 336 | |||
| XM_017029730.1 | XP_016885219.1 | 4 | 333 | |||
| XM_017029732.1 | XP_016885221.1 | 4 | 299 | |||
| XM_017029733.1 | XP_016885222.1 | 4 | 338 | |||
| GenBank mRNA | AK295054.1 | - | 4 | 268 | ||
| AK308986.1 | - | 4 | 303 | |||
| AY971607.1 | - | 4 | 320 | |||
| DA195605.1 | - | 4 | 336 | |||
| DC317540.1 | - | 4 | 320 | |||

KRBOX4
Genomic Map