siRNA Details
| Gene Symbol: OR5J2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005492.1 | NP_001005492.1 | 1 | 548 | ||
| GenBank mRNA | X64974.1 | - | 1 | 174 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR5J2| Gene Symbol: OR5J2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005492.1 | NP_001005492.1 | 1 | 548 | ||
| GenBank mRNA | X64974.1 | - | 1 | 174 | ||