siRNA Details
| Gene Symbol: Clec4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005891.1 | NP_001005891.1 | 3 | 309 | ||
| XM_006237321.2 | XP_006237383.1 | 3 | 518 | |||
| GenBank mRNA | AY397671.1 | - | 3 | 309 | ||
| AY494062.1 | - | 3 | 310 | |||
| FQ230051.1 | - | 3 | 404 | |||
| FQ234061.1 | - | 3 | 434 | |||

Clec4a3
Genomic Map