siRNA Details
| Gene Symbol: Clec4b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005896.1 | NP_001005896.1 | 6 | 705 | ||
| GenBank mRNA | AY365132.1 | - | 6 | 705 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec4b2| Gene Symbol: Clec4b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005896.1 | NP_001005896.1 | 6 | 705 | ||
| GenBank mRNA | AY365132.1 | - | 6 | 705 | ||