siRNA Details
| Gene Symbol: JMJD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005920.2 | NP_001005920.2 | 9 | 1688 | ||
| NM_001323918.1 | NP_001310847.1 | 9 | 1690 | |||
| NM_001323919.1 | NP_001310848.1 | 8 | 1560 | |||
| NM_001323920.1 | NP_001310849.1 | 8 | 1605 | |||
| NM_001323922.1 | NP_001310851.1 | 8 | 1600 | |||
| NR_136650.1 | 8 | 1778 | ||||
| NR_136651.1 | 8 | 1783 | ||||
| NR_136652.1 | 7 | 1693 | ||||
| GenBank mRNA | AF370420.1 | - | 9 | 1571 | ||
| BC073785.1 | - | 8 | 1659 | |||
| BC094850.1 | - | 6 | 1305 | |||

JMJD8
Genomic Map