siRNA Details
| Gene Symbol: JMJD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005920.2 | NP_001005920.2 | 4 | 380 | ||
| NM_001323918.1 | NP_001310847.1 | 4 | 387 | |||
| NM_001323919.1 | NP_001310848.1 | 4 | 387 | |||
| NM_001323920.1 | NP_001310849.1 | 3 | 297 | |||
| NM_001323922.1 | NP_001310851.1 | 3 | 297 | |||
| NR_136650.1 | 4 | 387 | ||||
| NR_136651.1 | 4 | 387 | ||||
| NR_136652.1 | 3 | 297 | ||||
| GenBank mRNA | AF370420.1 | - | 4 | 264 | ||
| AL524336.3 | - | 4 | 271 | |||
| AL524922.3 | - | 4 | 274 | |||
| BC073785.1 | - | 3 | 352 | |||
| BC137100.1 | - | 4 | 346 | 9021097 | ||
| BC137101.1 | - | 4 | 346 | 9021098 | ||
| BG826580.1 | - | 2 | 90 | |||
| BI463867.1 | - | 4 | 385 | |||
| BI767435.1 | - | 3 | 200 | |||
| BX335015.2 | - | 4 | 267 | |||
| CD674105.1 | - | 4 | 287 | |||
| CN371155.1 | - | 1 | 26 | |||
| DQ074695.1 | - | 3 | 170 | |||
| HY023676.1 | - | 3 | 285 | |||

JMJD8
Genomic Map