siRNA Details
| Gene Symbol: Olr1362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006598.1 | NP_001006598.1 | 1 | 408 | ||
| GenBank mRNA | AF091566.1 | - | 1 | 219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1362| Gene Symbol: Olr1362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006598.1 | NP_001006598.1 | 1 | 408 | ||
| GenBank mRNA | AF091566.1 | - | 1 | 219 | ||