siRNA Details
| Gene Symbol: FAM230C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027278.1 | 2 | 157 | |||
| GenBank mRNA | AK097777.1 | - | 2 | 103 | ||
| Gene Symbol: LOC388882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110539.1 | 2 | 144 | |||
| GenBank mRNA | BC036910.1 | - | 2 | 144 | 5164270 | |

FAM230C
Genomic Map