siRNA Details
Gene Symbol: Olr1493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006609.1 | NP_001006610.1 | 604 | |||
GenBank mRNA | AF091572.1 | - | 1 | 421 | ||
M64387.1 | - | 604 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr1493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006609.1 | NP_001006610.1 | 604 | |||
GenBank mRNA | AF091572.1 | - | 1 | 421 | ||
M64387.1 | - | 604 |