siRNA Details
| Gene Symbol: D8Ertd738e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007571.2 | NP_001007572.1 | 1 | 63 | ||
| GenBank mRNA | AK146163.1 | - | 1 | 21 | ||
| AK167785.1 | - | 1 | 22 | |||
| BC024662.1 | - | 1 | 12 | 4205912 | ||
| CF740495.1 | - | 1 | 63 | |||

D8Ertd738e
Genomic Map