siRNA Details
| Gene Symbol: Cmtm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007802.3 | NP_001007803.1 | 3, 4 | 356 | ||
| XM_006243969.2 | XP_006244031.1 | 2, 3 | 378 | |||
| GenBank mRNA | AY325262.1 | - | 3, 4 | 356 | ||
| AY387088.1 | - | 2, 3 | 316 | |||
| BC107930.1 | - | 2, 3 | 349 | 7130158 | ||

Cmtm6
Genomic Map