siRNA Details
| Gene Symbol: 1700001J03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008547.1 | NP_001008547.1 | 5 | 742 | ||
| GenBank mRNA | AK005597.1 | - | 5 | 739 | ||
| BC048528.1 | - | 5 | 742 | 6774214 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700001J03Rik| Gene Symbol: 1700001J03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008547.1 | NP_001008547.1 | 5 | 742 | ||
| GenBank mRNA | AK005597.1 | - | 5 | 739 | ||
| BC048528.1 | - | 5 | 742 | 6774214 | ||