siRNA Details
| Gene Symbol: ZNF121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008727.3 | NP_001008727.1 | 3 | 218 | ||
| NM_001308269.1 | NP_001295198.1 | 3 | 218 | |||
| XM_017027239.1 | XP_016882728.1 | 4 | 392 | |||
| GenBank mRNA | AU253046.1 | - | 3 | 218 | ||
| CN360954.1 | - | 2 | 164 | |||

ZNF121
Genomic Map