siRNA Details
| Gene Symbol: ZNF121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008727.3 | NP_001008727.1 | 4 | 490 | ||
| NM_001308269.1 | NP_001295198.1 | 5 | 548 | |||
| XM_017027239.1 | XP_016882728.1 | 5 | 664 | |||
| GenBank mRNA | AK297919.1 | - | 5 | 547 | ||
| AU253046.1 | - | 4 | 490 | |||
| CN360954.1 | - | 3 | 436 | |||
| DC424100.1 | - | 5 | 547 | |||

ZNF121
Genomic Map