siRNA Details
| Gene Symbol: RAB19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008749.2 | NP_001008749.2 | 2 | 386 | ||
| XM_005249996.3 | XP_005250053.1 | 2 | 669 | |||
| XM_017012207.1 | XP_016867696.1 | 2 | 582 | |||
| GenBank mRNA | AF091033.1 | - | 1 | 104 | ||
| BC140796.1 | - | 1 | 256 | 9021664 | ||
| BC171745.1 | - | 1 | 256 | |||
| DN996121.1 | - | 2 | 360 | |||

RAB19
Genomic Map