siRNA Details
| Gene Symbol: RASIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017805.2 | NP_060275.2 | 8 | 2287 | ||
| XM_011527053.2 | XP_011525355.2 | 8 | 2407 | |||
| XM_011527054.2 | XP_011525356.2 | 8 | 2407 | |||
| XM_017026914.1 | XP_016882403.1 | 8 | 2407 | |||
| GenBank mRNA | AK000408.1 | - | 4 | 402 | ||
| AY378097.1 | - | 8 | 2152 | |||
| BC021860.1 | - | 4 | 417 | |||
| BC028614.1 | - | 8 | 2159 | 4815880 | ||

RASIP1
Genomic Map