siRNA Details
| Gene Symbol: Vom1r59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008944.1 | NP_001008944.1 | 1 | 379 | ||
| GenBank mRNA | AY510282.1 | - | 1 | 379 | ||
| Gene Symbol: LOC103691064 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759093.1 | XP_008757315.1 | 61 | |||

Vom1r59
Genomic Map