siRNA Details
| Gene Symbol: Qprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009646.1 | NP_001009646.1 | 3 | 646 | ||
| XM_006230246.2 | XP_006230308.1 | 3 | 817 | |||
| XM_008759877.1 | XP_008758099.1 | 3 | 1122 | |||
| XM_008759878.1 | XP_008758100.1 | 4 | 909 | |||
| GenBank mRNA | BC088177.1 | - | 3 | 646 | 7323872 | |
| FQ209866.1 | - | 3 | 640 | |||

Qprt
Genomic Map