siRNA Details
| Gene Symbol: TMEM161A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017814.2 | NP_060284.1 | 5 | 444 | ||
| XM_017026916.1 | XP_016882405.1 | 4, 5 | 340 | |||
| GenBank mRNA | AK000429.1 | - | 5 | 401 | ||
| AK225424.1 | - | 5 | 397 | |||
| AY358088.1 | - | 5 | 428 | |||
| BC005210.1 | - | 5 | 404 | 4041226 | ||
| BC016478.2 | - | 4 | 368 | |||
| DA268563.1 | - | 5 | 444 | |||

TMEM161A
Genomic Map