siRNA Details
| Gene Symbol: Tmem140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009709.1 | NP_001009709.1 | 2 | 875 | ||
| GenBank mRNA | BC088293.1 | - | 2 | 875 | 7322541 | |
| FQ225337.1 | - | 2 | 993 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem140| Gene Symbol: Tmem140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009709.1 | NP_001009709.1 | 2 | 875 | ||
| GenBank mRNA | BC088293.1 | - | 2 | 875 | 7322541 | |
| FQ225337.1 | - | 2 | 993 | |||