siRNA Details
| Gene Symbol: Lrrc41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009710.1 | NP_001009710.1 | 4, 5 | 1515 | ||
| XM_006238682.2 | XP_006238744.1 | 4, 5 | 1900 | |||
| GenBank mRNA | BC087037.1 | - | 4, 5 | 1515 | 7133544 | |

Lrrc41
Genomic Map