siRNA Details
| Gene Symbol: RGD1304567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009711.1 | NP_001009711.1 | 3 | 243 | ||
| GenBank mRNA | BC087640.1 | - | 3 | 243 | 7308844 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1304567| Gene Symbol: RGD1304567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009711.1 | NP_001009711.1 | 3 | 243 | ||
| GenBank mRNA | BC087640.1 | - | 3 | 243 | 7308844 | |