siRNA Details
Gene Symbol: Lrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009717.1 | NP_001009717.1 | 2 | 1266 | ||
GenBank mRNA | BC088434.1 | - | 2 | 1266 | 7124833 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009717.1 | NP_001009717.1 | 2 | 1266 | ||
GenBank mRNA | BC088434.1 | - | 2 | 1266 | 7124833 |