siRNA Details
| Gene Symbol: Spetex-2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009976.1 | NP_001009976.1 | 1 | 45 | ||
| GenBank mRNA | AB180076.1 | - | 1 | 45 | ||
| Gene Symbol: LOC691414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766795.1 | XP_008765017.1 | 1 | 142 | ||

Spetex-2A
Genomic Map