siRNA Details
| Gene Symbol: C10orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009997.2 | NP_001009997.2 | 1 | 283 | ||
| GenBank mRNA | BC037252.1 | - | 1 | 249 | 5163660 | |
| BC043281.1 | - | 1 | 734 | |||
| BI826342.1 | - | 1 | 282 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C10orf62| Gene Symbol: C10orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009997.2 | NP_001009997.2 | 1 | 283 | ||
| GenBank mRNA | BC037252.1 | - | 1 | 249 | 5163660 | |
| BC043281.1 | - | 1 | 734 | |||
| BI826342.1 | - | 1 | 282 | |||