siRNA Details
| Gene Symbol: ZIK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010879.3 | NP_001010879.2 | 4 | 2335 | ||
| NM_001321145.1 | NP_001308074.1 | 3 | 2296 | |||
| NM_001321146.1 | NP_001308075.1 | 3 | 2519 | |||
| NM_001321147.1 | NP_001308076.1 | 2 | 2169 | |||
| XM_011526761.1 | XP_011525063.1 | 4 | 2627 | |||
| XM_011526762.1 | XP_011525064.1 | 4 | 2892 | |||
| XM_011526763.1 | XP_011525065.1 | 3 | 2588 | |||
| XM_011526764.1 | XP_011525066.1 | 4 | 1970 | |||
| XM_011526765.2 | XP_011525067.1 | 2 | 2726 | |||
| XM_011526766.2 | XP_011525068.1 | 3 | 1909 | |||
| XM_011526767.2 | XP_011525069.1 | 2 | 2626 | |||
| GenBank mRNA | AK092538.1 | - | 4 | 2169 | ||
| AK128340.1 | - | 3 | 2130 | |||
| BC103957.1 | - | 2 | 1885 | 40011908 | ||
| BC103958.1 | - | 4 | 2051 | 40011910 | ||
| BC103959.2 | - | 3 | 2235 | 40011911 | ||

ZIK1
Genomic Map