siRNA Details
| Gene Symbol: C6orf222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010903.4 | NP_001010903.3 | 11, 12 | 2106 | ||
| XM_011514596.2 | XP_011512898.1 | 11, 12 | 2959 | |||
| XM_011514597.2 | XP_011512899.1 | 11, 12 | 2233 | |||
| XM_011514598.2 | XP_011512900.1 | 10, 11 | 1616 | |||
| GenBank mRNA | BC140873.1 | - | 11, 12 | 2029 | 9021741 | |
| BX648558.1 | - | 11, 12 | 2107 | |||

C6orf222
Genomic Map