siRNA Details
| Gene Symbol: C6orf222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010903.4 | NP_001010903.3 | 12 | 2875 | ||
| XM_011514596.2 | XP_011512898.1 | 12 | 3728 | |||
| XM_011514597.2 | XP_011512899.1 | 12 | 3002 | |||
| XM_011514598.2 | XP_011512900.1 | 11 | 2385 | |||
| GenBank mRNA | BX648558.1 | - | 12 | 2876 | ||

C6orf222
Genomic Map