siRNA Details
| Gene Symbol: C9orf153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276366.1 | NP_001263295.1 | 1 | 90 | ||
| NM_001276367.1 | NP_001263296.1 | 1 | 90 | |||
| GenBank mRNA | BG772848.1 | - | 1 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9orf153| Gene Symbol: C9orf153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276366.1 | NP_001263295.1 | 1 | 90 | ||
| NM_001276367.1 | NP_001263296.1 | 1 | 90 | |||
| GenBank mRNA | BG772848.1 | - | 1 | 95 | ||