siRNA Details
| Gene Symbol: MARCH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017824.4 | NP_060294.1 | 2 | 441 | ||
| XM_011539886.2 | XP_011538188.1 | 2 | 433 | |||
| GenBank mRNA | AB191202.1 | - | 2 | 197 | ||
| AK000452.1 | - | 2 | 443 | |||
| AK294799.1 | - | 2 | 515 | |||
| AK309175.1 | - | 2 | 441 | |||
| BC015480.1 | - | 2 | 401 | 3905766 | ||

MARCH5
Genomic Map