siRNA Details
| Gene Symbol: LINC00200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015376.2 | 3 | 480 | |||
| GenBank mRNA | AK097673.1 | - | 3 | 430 | ||
| BC133038.1 | - | 3 | 480 | |||
| BC133040.1 | - | 3 | 432 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00200| Gene Symbol: LINC00200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015376.2 | 3 | 480 | |||
| GenBank mRNA | AK097673.1 | - | 3 | 430 | ||
| BC133038.1 | - | 3 | 480 | |||
| BC133040.1 | - | 3 | 432 | |||