siRNA Details
Gene Symbol: C10orf120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010912.2 | NP_001010912.1 | 3 | 812 | ||
GenBank mRNA | BC065903.1 | - | 3 | 816 | ||
BC140922.1 | - | 3 | 804 | 9056734 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C10orf120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010912.2 | NP_001010912.1 | 3 | 812 | ||
GenBank mRNA | BC065903.1 | - | 3 | 816 | ||
BC140922.1 | - | 3 | 804 | 9056734 |