siRNA Details
| Gene Symbol: C10orf120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010912.2 | NP_001010912.1 | 3 | 575 | ||
| GenBank mRNA | BC065903.1 | - | 3 | 579 | ||
| BC140922.1 | - | 3 | 567 | 9056734 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C10orf120| Gene Symbol: C10orf120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010912.2 | NP_001010912.1 | 3 | 575 | ||
| GenBank mRNA | BC065903.1 | - | 3 | 579 | ||
| BC140922.1 | - | 3 | 567 | 9056734 | ||