siRNA Details
| Gene Symbol: SARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145901.1 | NP_001139373.1 | 1 | 235 | ||
| NM_017827.3 | NP_060297.1 | 1 | 235 | |||
| GenBank mRNA | AB029948.1 | - | 1 | 235 | ||
| AK000457.1 | - | 1 | 102 | |||
| AK293414.1 | - | 1 | 100 | |||
| AK296152.1 | - | 1 | 99 | |||
| AK296170.1 | - | 1 | 102 | |||
| BC042912.1 | - | 1 | 89 | 5478759 | ||

SARS2
Genomic Map