siRNA Details
| Gene Symbol: Zc3h15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010963.1 | NP_001010963.1 | 2 | 192 | ||
| GenBank mRNA | AY377983.1 | - | 2 | 148 | ||
| BC088438.1 | - | 2 | 192 | 7126102 | ||
| FQ232115.1 | - | 2 | 271 | |||
| FQ233565.1 | - | 2 | 294 | |||

Zc3h15
Genomic Map