siRNA Details
| Gene Symbol: Zc3h15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010963.1 | NP_001010963.1 | 9 | 1111 | ||
| GenBank mRNA | AY377983.1 | - | 9 | 1067 | ||
| BC088438.1 | - | 9 | 1111 | 7126102 | ||
| FQ212631.1 | - | 4 | 399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h15| Gene Symbol: Zc3h15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010963.1 | NP_001010963.1 | 9 | 1111 | ||
| GenBank mRNA | AY377983.1 | - | 9 | 1067 | ||
| BC088438.1 | - | 9 | 1111 | 7126102 | ||
| FQ212631.1 | - | 4 | 399 | |||