siRNA Details
| Gene Symbol: C1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010979.2 | NP_001010979.1 | 4 | 405 | ||
| GenBank mRNA | BC127710.1 | - | 4 | 404 | 40127224 | |
| HY025946.1 | - | 4 | 405 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1orf189| Gene Symbol: C1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010979.2 | NP_001010979.1 | 4 | 405 | ||
| GenBank mRNA | BC127710.1 | - | 4 | 404 | 40127224 | |
| HY025946.1 | - | 4 | 405 | |||