siRNA Details
| Gene Symbol: IFIT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010987.2 | NP_001010987.1 | 2 | 1787 | ||
| GenBank mRNA | BC150189.1 | - | 2 | 1769 | 40148511 | |
| BX647300.1 | - | 2 | 1787 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IFIT1B| Gene Symbol: IFIT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010987.2 | NP_001010987.1 | 2 | 1787 | ||
| GenBank mRNA | BC150189.1 | - | 2 | 1769 | 40148511 | |
| BX647300.1 | - | 2 | 1787 | |||