siRNA Details
| Gene Symbol: SLC9A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011552.3 | NP_001011552.2 | 12 | 3627 | ||
| XM_011511158.1 | XP_011509460.1 | 12 | 3561 | |||
| GenBank mRNA | AL833048.1 | - | 1 | 1150 | ||
| CR627411.1 | - | 12 | 3628 | |||

SLC9A4
Genomic Map