siRNA Details
| Gene Symbol: XKR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011720.1 | NP_001011720.1 | 3 | 696 | ||
| NM_001287258.1 | NP_001274187.1 | 3 | 696 | |||
| NM_001287259.1 | NP_001274188.1 | 4 | 806 | |||
| NM_001287260.1 | NP_001274189.1 | 3 | 550 | |||
| XM_011517524.2 | XP_011515826.1 | 3 | 679 | |||
| XM_011517525.2 | XP_011515827.1 | 3 | 541 | |||
| XM_011517526.2 | XP_011515828.1 | 3 | 691 | |||
| XM_011517527.2 | XP_011515829.1 | 3 | 691 | |||
| XM_017013404.1 | XP_016868893.1 | 2 | 913 | |||
| XM_017013405.1 | XP_016868894.1 | 3 | 691 | |||
| GenBank mRNA | AY534247.1 | - | 3 | 696 | ||
| BC137104.1 | - | 3 | 614 | 9021101 | ||
| BC137105.1 | - | 3 | 550 | 9021102 | ||
| BC151845.1 | - | 4 | 773 | |||

XKR9
Genomic Map