siRNA Details
| Gene Symbol: XKR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011720.1 | NP_001011720.1 | 5 | 2372 | ||
| NM_001287258.1 | NP_001274187.1 | 6 | 2490 | |||
| NM_001287259.1 | NP_001274188.1 | 6 | 2482 | |||
| NM_001287260.1 | NP_001274189.1 | 5 | 2226 | |||
| GenBank mRNA | AY534247.1 | - | 5 | 2372 | ||

XKR9
Genomic Map