siRNA Details
Gene Symbol: Olfr212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011800.2 | NP_001011800.1 | 2 | 1206 | ||
GenBank mRNA | AK029829.1 | - | 2 | 1208 | ||
BC141000.1 | - | 2 | 1015 | 9055612 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011800.2 | NP_001011800.1 | 2 | 1206 | ||
GenBank mRNA | AK029829.1 | - | 2 | 1208 | ||
BC141000.1 | - | 2 | 1015 | 9055612 |