siRNA Details
| Gene Symbol: Xkr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011873.2 | NP_001011873.1 | 3 | 762 | ||
| GenBank mRNA | AY534255.1 | - | 3 | 762 | ||
| BC147699.1 | - | 2 | 368 | 9056672 | ||
| BC158121.1 | - | 2 | 623 | 9088194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xkr9| Gene Symbol: Xkr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011873.2 | NP_001011873.1 | 3 | 762 | ||
| GenBank mRNA | AY534255.1 | - | 3 | 762 | ||
| BC147699.1 | - | 2 | 368 | 9056672 | ||
| BC158121.1 | - | 2 | 623 | 9088194 | ||