siRNA Details
| Gene Symbol: Sf3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011986.1 | NP_001011986.1 | 9 | 1574 | ||
| XM_006240885.2 | XP_006240947.1 | 8 | 1531 | |||
| XM_006240886.2 | XP_006240948.1 | 9 | 1828 | |||
| GenBank mRNA | BC079320.1 | - | 9 | 1574 | 7135238 | |

Sf3a2
Genomic Map