siRNA Details
| Gene Symbol: Upf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012159.1 | NP_001012159.1 | 9 | 1467 | ||
| GenBank mRNA | BC088463.1 | - | 9 | 1467 | 7127783 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Upf3a| Gene Symbol: Upf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012159.1 | NP_001012159.1 | 9 | 1467 | ||
| GenBank mRNA | BC088463.1 | - | 9 | 1467 | 7127783 | |